Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ Wnt Antagonist III, Box5
Shop All MilliporeSigma Products
Click to view available options
Quantity:
10 mg
Description
- Packaged Under Inert Gas
- Peptide Sequence: t-Boc-NH-Met-Asp-Gly-Cys-Glu-Leu-CO2H
Specifications
Specifications
| Color | White |
| Description | 90% by HPLC |
| Molecular Formula | C30H50N6O13S2 |
| Formula Weight | 766.9g/mol |
| Solubility | DMSO or H2O |
| Content And Storage | Protect from light |
| Form | Lyophilized |
| Quantity | 10 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction