Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MilliporeSigma™ Calbiochem™ (Z-LL)2 Ketone
Click to view available options
Quantity:
5 mg
Description
- Packaged Under Inert Gas
- Peptide Sequence: Z-Leu-Leu-NH-CH2-CO-CH2-NH-Leu-Leu-Z

Specifications
Specifications
Color | Off-white |
Description | single spot by TLC |
Target Proteins | Processing of the p-Prl signal peptide |
Molecular Formula | C43H64N6O9 |
Formula Weight | 809g/mol |
Solubility | DMSO |
Form | Lyophilized |
Quantity | 5 mg |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction