Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CALCA Mouse anti-Human, Clone: 406CT21.4.3, Abnova™

Catalog No. 89933709 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-933-709 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-933-709 Supplier Abnova Corporation Supplier No. MAB12852
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against synthetic peptide of human CALCA.

Calcitonin is a peptide hormone synthesized by the parafollicular cells of the thyroid. It causes reduction in serum calcium, an effect opposite to that of parathyroid hormone (PTH; MIM 168450).[supplied by OMIM]

Specifications

Antigen CALCA
Applications Immunohistochemistry (PFA fixed), Western Blot
Classification Monoclonal
Clone 406CT21.4.3
Conjugate Unconjugated
Description Mouse monoclonal antibody raised against synthetic peptide of human CALCA.
Dilution Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:100) Western Blot (1:100-1:250) The optimal working dilution should be determined by the end user.
Formulation In PBS (0.09% sodium azide).
Gene Alias CALC1/CGRP/CGRP-I/CGRP1/CT/KC/MGC126648
Gene Symbols CALCA
Host Species Mouse
Immunogen A synthetic peptide (conjugated with KLH) corresponding to amino acids 56-84 of human CALCA.
Purification Method Euglobulin precipitation
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 796
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Form Liquid
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.