Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CALCB Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA596934
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA596934 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA596934 Supplier Invitrogen™ Supplier No. PA596934
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MGFRKFSPFL ALSILVLYQA GSLQAAPFRS ALESSPDPAT LSKEDARLLL AALVQDYVQM KASELKQEQE TQGSSSAAQK RACNTATCVT HRLAGLLSRS GGMVKSNFVP TNVGSKAFGR RRRDLQA; Positive Samples: 293T, HeLa.

CGRP induces vasodilation. It dilates a variety of vessels including the coronary, cerebral and systemic vasculature. Its abundance in the CNS also points toward a neurotransmitter or neuromodulator role.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CALCB
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.64 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene CALCB
Gene Accession No. P10092, P10093, Q99MP3
Gene Alias Beta-CGRP; beta-type CGRP; CALC2; Calcb; calcitonin 2; calcitonin gene-related peptide 2; Calcitonin gene-related peptide II; calcitonin related polypeptide beta; calcitonin-related polypeptide beta; calcitonin-related polypeptide, beta; CGRP2; CGRP-II
Gene Symbols CALCB
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-127 of human CALCB (NP_0007191).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116903, 171519, 797
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.