Learn More
R&D Systems™ Calcein AM
SDP
Catalog No. 489201001 Shop All Novus Products
Change view
Click to view available options
Quantity:
50 μg
Catalog No. Quantity
48-920-1001 50 μg
1 options
This item is not returnable. View return policy
Catalog No. 48-920-1001 Supplier R&D Systems™ Supplier No. 489201001
Add to Cart
Add to Cart
This item is not returnable. View return policy
R&D Systems™ Calcein AM provides a simple, rapid and accurate method to measure cell viability and/or cytotoxicity.
  • Use a manual defrost freezer and avoid repeated freeze-thaw cycles.
  • Do not use past expiration date.
  • The hydrolysis of Calcein AM by intracellular esterases produces calcein (structure B), a hydrophilic, strongly fluorescent compound that is well-retained in the cell cytoplasm.
  • Cells grown in black-walled plates can be stained and quantified in less than two hours.
  • Non-fluorescent, hydrophilic compound that easily permeates intact, live cells
Specifications
Content And Storage Store the unopened product at -20 to -70° C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
Detection Method Fluorometric
Product Line R&D Systems
Quantity 50 μg
Druggable Target Cell-based Assay