Learn More
Calcein AM Solution, Sigma-Aldrich
Catalog No. 01259012 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
01-259-012 100 μL
1 options
Catalog No. 01-259-012 Supplier MilliporeSigma Supelco Supplier No. C1359100UL
Add to Cart
Add to Cart
Calcein AM is a membrane permeable nonfluorescent molecule. It is converted to fluorescent calcein by intracellular esterases. Calcein has limited membrane permeability. Calcein AM is also a substrate for p-glycoprotein (Pgp). Presence of Pgp results in efflux of calcein AM and thereby decreases cellular fluorescence. On the other hand, presence of Pgp inhibitor increases cellular entry of calcein AM and thereby increases cellular fluorescence. Calcein AM plays an important role in early drug discovery.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 4 mM
Product Type Stain