Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Calcein AM Viability Dye (UltraPure Grade)

Catalog No. 5016953
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
20 x 50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-169-53 20 x 50 μg
50-169-52 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-169-53 Supplier Invitrogen™ Supplier No. 65085378
  • $346.00
    $335.65 / Each of 1

    Save $10.35

    3% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

A membrane-permeable live-cell labeling dye. Upon entering the cell, intracellular esterases cleave the acetoxymethyl (AM) ester group, yielding the membrane-impermeable Calcein fluorescent dye.

  • Upon entering the cell, intracellular esterases cleave the acetoxymethyl (AM) ester group, yielding the membrane-impermeable Calcein fluorescent dye
  • Apoptotic and dead cells with compromised cell membranes do not retain Calcein
  • Calcein is optimally excited at 495 nm and has a peak emission of 515 nm
  • For flow cytometric analysis, it can be excited off the blue laser line (488 nm) and detected using filters for FITC/Alexa Fluor™ 488 (530/30)
  • For multicolor flow cytometry, concentrations in the range of 5 to 20 nM are recommended to ensure that compensation can be performed successfully
  • Co-staining with Annexin V or 7-AAD is recommended to allow the greatest resolution between live and dead/apoptotic cells
  • If single-color analysis is desired, improved resolution between live and dead/apoptotic cells may be obtained by using either Calcein Blue AM (cat. 65-0855) or Calcein Violet 450 AM (cat. 65-0854)
  • Calcein AM should be reconstituted in high-quality, freshly opened DMSO
  • Once reconstituted, it should be stored at -20°C with dessicant and used within a short period of time (avoid freeze-thawing)
  • Purity: >95% (determined by HPLC)
TRUSTED_SUSTAINABILITY

Specifications

Color Green
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Dye
Product Line eBioscience
Quantity 20 x 50 μg
Emission Wavelength 495/515
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.