Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

R&D Systems™ Calcein AM
GREENER_CHOICESDP

Catalog No. 51191 Shop All Novus Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
51-191 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 51-191 Supplier R&D Systems™ Supplier No. 5119/1

Customers require a valid business account associated to their profile to access pricing and ordering options.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Cell permeable compound; hydrolyzed to become fluorescent in living cells

Calcein AM is a cell permeable, non-fluorescent compound for monitoring cell viability, chemotaxis, cell adhesion and multidrug resistance; hydrolyzed by intracellular esterases to become fluorescent calcein in living cells. .

Specifications

CAS 890090-35-4
Quantity 1 mg
Product Line Tocris
Product Type Calcein AM
Purity 95%
Molecular Weight (g/mol) M.W. (Observed): 994.86
Inhibitors Calcein AM

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.