Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ eBioscience™ Calcein Blue AM Viability Dye

Catalog No. 5016955
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-169-55 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-169-55 Supplier Invitrogen™ Supplier No. 65085539
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Membrane-permeable live-cell labeling dye. Upon entering the cell, intracellular esterases cleave the acetoxymethyl (AM) ester group, yielding the membrane-impermeable Calcein Blue fluorescent dye. Apoptotic and dead cells with compromised cell membranes do not retain Calcein Blue.

  • Molecular weight: 465.41g/mol
  • Peak excitation: 360nm
  • Peak emission: 445nm
  • Reconstitute in high-quality, freshly opened DMSO
  • Once reconstituted, store at −20°C with dessicant and use within a short period of time. Avoid freeze-thawing
  • Calcein Blue is optimally excited by the UV laser line (360nm), but can also be excited by the violet laser line (405nm)
  • For fluorescent microscopy, detect using the appropriate filter sets
  • Calcein Blue AM is itself weakly fluorescent, and an additional wash step may be necessary to reduce background fluorescence
  • Co-staining with Annexin V or 7-AAD is recommended to allow the greatest resolution between live and dead/apoptotic cells
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
Detection Method Fluorescence
For Use With (Application) Viability Assay
For Use With (Equipment) Fluorescence Microscope, Flow Cytometer
Product Type Dye
Product Line eBioscience
Quantity 1 mg
Emission Wavelength 360/450

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.