Learn More
Description
Calcein has been used:
- in whole cell staining
- in histomorphometry and micro-computed tomography
- to label active bone-forming surfaces
Specifications
Specifications
| Product Type | Fluorescence Reagent |
| Molecular Formula | C30H26N2O13 |
| InChI Key | DEGAKNSWVGKMLS-UHFFFAOYSA-N |
| Quantity | 10 g |
| SMILES | OC(=O)CN(CC(O)=O)CC1=C(O)C=C2OC3=CC(O)=C(CN(CC(O)=O)CC(O)=O)C=C3C3(OC(=O)C4=CC=CC=C34)C2=C1 |
| IUPAC Name | 2-{[(7'-{[bis(carboxymethyl)amino]methyl}-3',6'-dihydroxy-3-oxo-3H-spiro[2-benzofuran-1,9'-xanthen]-2'-yl)methyl](carboxymethyl)amino}acetic acid |
| Molecular Weight (g/mol) | 622.54, 622.53 g/mol |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.