Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcein, Sigma-Aldrich

Catalog No. 01258998 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 g
10 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-998 5 g
01-258-854 10 g
01-258-996 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-258-998 Supplier MilliporeSigma Supelco Supplier No. C08755G
Add to Cart
Add to Cart

Used for the fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium.

Calcein has been used:

  • in whole cell staining
  • in histomorphometry and micro-computed tomography
  • to label active bone-forming surfaces
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 154071-48-4

Specifications

Product Type Fluorescence Reagent
Quantity 5 g
Molecular Weight (g/mol) 622.53 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.