Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcineurin A Antibody (BF1.1), DyLight 550, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308394R
Description
Calcineurin A Monoclonal antibody specifically detects Calcineurin A in Human samples. It is validated for Western Blot, ELISASpecifications
Calcineurin A | |
Monoclonal | |
DyLight 550 | |
calcineurin A alpha, Calmodulin-dependent calcineurin A subunit alpha isoform, CALN, CALNAcatalytic subunit, alpha isoform, CAM-PRP catalytic subunit, CNA, CNA1CALNA1, EC 3.1.3.16, PPP2BCCN1, protein phosphatase 2B, catalytic subunit, alpha isoform, protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform(calcineurin A alpha), protein phosphatase 3, catalytic subunit, alpha isozyme, serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform | |
Recombinant full-length Calcineurin A protein (Uniprot: Q08209) | |
100 μg | |
Adaptive Immunity, Immunology, Protein Phosphatase, Wnt Signaling Pathway | |
5530 | |
Store at 4°C in the dark. | |
IgG1 κ |
Western Blot, ELISA | |
BF1.1 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction