Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcineurin A Mouse anti-Human, Clone: CALNA/2353, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$268.00

Specifications

Antigen Calcineurin A
Clone CALNA/2353
Concentration 0.2 mg/ml
Dilution Immunohistochemistry-Paraffin 1-2 ug/ml
Applications Immunohistochemistry (Paraffin)
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB122790
SDP
View Documents
Novus Biologicals
NBP30765820UG
20 μg
Each of 1 for $268.00
Only null left
Add to Cart
 
Description

Description

Calcineurin A Monoclonal specifically detects Calcineurin A in Human samples. It is validated for Immunohistochemistry-Paraffin.
Specifications

Specifications

Calcineurin A
0.2 mg/ml
Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Adaptive Immunity, Immunology, Protein Phosphatase, Wnt Signaling Pathway
10 mM PBS with 0.05% BSA
5530
IgG1 κ
Protein A or G purified
CALNA/2353
Immunohistochemistry-Paraffin 1-2 ug/ml
Monoclonal
Purified
RUO
Human
calcineurin A alpha, Calmodulin-dependent calcineurin A subunit alpha isoform, CALN, CALNAcatalytic subunit, alpha isoform, CAM-PRP catalytic subunit, CNA, CNA1CALNA1, EC 3.1.3.16, PPP2BCCN1, protein phosphatase 2B, catalytic subunit, alpha isoform, protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform(calcineurin A alpha), protein phosphatase 3, catalytic subunit, alpha isozyme, serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform
Recombinant full-length human Calcineurin A protein
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.