Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calcineurin A Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA529255
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA529255 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA529255 Supplier Invitrogen™ Supplier No. PA529255
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: human PPP3CA-transfected 293T, mouse brain, Rat brain. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (93%), Xenopus laevis (98%), Dog (100%), Pig (99%), Rabbit (100%), Rhesus Monkey (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Calcineurin plays an important role in the T-cell receptor-mediated signal transduction pathway. The protein encoded by this gene binds specifically to the activated form of calcineurin and inhibits calcineurin-mediated signal transduction. The encoded protein is found in the nucleus and contains a leucine zipper domain as well as several PEST motifs, sequences which confer targeted degradation to those proteins which contain them. Alternative splicing results in multiple transcript variants encoding two different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcineurin A
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.77 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene PPP3CA
Gene Accession No. P63328, P63329, Q08209
Gene Alias 2900074D19Rik; Ca2+/calmodulin-regulated protein phosphatase; calcineurin; calcineurin A alpha; Calcineurin A1; calcineurin catalytic subunit; calcineurin catalytic subunit delta isoform; Calcineurin subunit A alpha; CalcineurinA; calmodulin-dependent calcineurin A subunit alpha isoform; caln; CALNA; CALNA 1; CALNA1; CAM-PRP catalytic subunit; CCN 1; ccn1; CN; CnA; CNA alpha; CNA1; PP2B alpha 1; PP2BA alpha; PP2CB; ppp2b; ppp3ca; ppp3ca.S; protein phosphatase; protein phosphatase 2B, catalytic subunit, alpha isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform; protein phosphatase 3 (formerly 2B), catalytic subunit, alpha isoform (calcineurin A alpha); protein phosphatase 3 catalytic subunit alpha; protein phosphatase 3 catalytic subunit alpha isozyme; protein phosphatase 3, catalytic subunit, alpha isoform; protein phosphatase 3, catalytic subunit, alpha isozyme; protein phosphatase 3, catalytic subunit, alpha isozyme S homeolog; protein phosphatase 3, catalytic subunit, gamma isozyme; protein phosphatase-2Ba; serine/threonine protein kinase; serine/threonine-protein phosphatase 2B catalytic subunit alpha isoform; XELAEV_18009418mg
Gene Symbols PPP3CA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Calcineurin A. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19055, 24674, 5530
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.