Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calcineurin B Monoclonal Antibody (CN-A1)
GREENER_CHOICE

Catalog No. PIMA549305
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA549305 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA549305 Supplier Invitrogen™ Supplier No. MA549305
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Adding 1 mL of PBS buffer will yield a concentration of 100 μg/mL. Positive Control - WB: rat brain tissue, mouse brain tissue. IHC: mouse brain tissue, rat brain tissue.

Calcineurin is a Ser/Thr, calcium and calmodulin-dependent protein phosphatase that plays an essential role in the T cell activation pathway. Calcineurin (also referred to as protein phosphatase 2B) is composed of two subunits; calcineurin A (CnA), a 60 kDa catalytic subunit which is highly homologous to protein phosphatases 1 and 2A; and a 19 kDa regulatory subunit, calcineurin B (CnB) which, like calmodulin, contains four EF-hand calcium binding sites. Calcineurin-mediated dephosphorylation of the nuclear factor of activated T-cells (NF-AT) is essential for NF-AT activation, nuclear translocation, and early gene expression in T-cells. The immunosuppressive drugs, cyclosporin A and FK506, when bound to the immunophilins, cyclophilin and FKBP respectively, inhibit dephosphorylation and activation of NF-AT by calcineurin. This inhibition is caused by the immunosuppressant/immunophilin complex simultaneously binding to both CnA and CnB which does not inhibit the phosphatase activity of CnA, but rather blocks access of the substrate to the catalytic domain of CnA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcineurin B
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CN-A1
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation Ascites, 1.2% sodium acetate with 2mg BSA and 0.01mg sodium azide
Gene PPP3R1
Gene Accession No. P63098, P63099, P63100, Q63810
Gene Alias ALNB1; calcineurin B, type I (19kDa); calcineurin subunit B type 1; CALNB1; CaNB1; Cna2; Cnb; Cnb1; MCIP1; PP2B beta 1; PPP3R1; protein phospatase 3, regulatory subunit B, alpha isoform (calcineurin B, type I); protein phospatase 3, regulatory subunit B, alpha isoform,type 1; protein phosphatase 2B regulatory subunit 1; protein phosphatase 2B regulatory subunit B alpha; protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), alpha isoform (calcineurin B, type I); protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, alpha isoform (calcineurin B, type I); protein phosphatase 3 (formerly 2B), regulatory subunit B, alpha isoform; protein phosphatase 3 regulatory subunit B alpha; Protein phosphatase 3 regulatory subunit B alpha isoform 1; protein phosphatase 3 regulatory subunit B, alpha; protein phosphatase 3, regulatory subunit B, alpha; protein phosphatase 3, regulatory subunit B, alpha isoform (calcineurin B, type I)
Gene Symbols PPP3R1
Host Species Mouse
Immunogen Bovine brain calcineurin.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19058, 282321, 29748, 5534
Target Species Human, Mouse, Rat, Bovine
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.