Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcineurin B Mouse anti-Human, Xenopus, Clone: BE2.1, Novus Biologicals™

Mouse Monoclonal Antibody
$268.00
Specifications
Antigen | Calcineurin B |
---|---|
Clone | BE2.1 |
Concentration | 0.2 mg/ml |
Dilution | Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml |
Applications | Western Blot, ELISA, Immunohistochemistry (Paraffin) |
Description
Calcineurin B Monoclonal specifically detects Calcineurin B in Human, Xenopus samples. It is validated for Western Blot, ELISA, Immunohistochemistry-Paraffin.Specifications
Calcineurin B | |
0.2 mg/ml | |
Western Blot, ELISA, Immunohistochemistry (Paraffin) | |
Unconjugated | |
Mouse | |
Protein Phosphatase, Signal Transduction, Wnt Signaling Pathway | |
10 mM PBS with 0.05% BSA | |
5534 | |
IgG2a κ | |
Protein A or G purified |
BE2.1 | |
Western Blot 1-2 ug/ml, ELISA, Immunohistochemistry-Paraffin 1-2 ug/ml | |
Monoclonal | |
Purified | |
RUO | |
Human, Xenopus species | |
calcineurin subunit B type 1, CNA2, CNB1, Protein phosphatase 2B regulatory subunit 1, protein phosphatase 2B regulatory subunit B alpha, protein phosphatase 3 (formerly 2B), regulatory subunit B (19kD), alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, 19kDa, alpha isoform(calcineurin B, type I), protein phosphatase 3 (formerly 2B), regulatory subunit B, alpha isoform, Protein phosphatase 3 regulatory subunit B alpha isoform 1, protein phosphatase 3, regulatory subunit B, alpha, type I (19kDa) | |
Recombinant full-length human Calcineurin B protein | |
Primary | |
Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title