Learn More
Description
Host Species: Rabbit
Isotype: IgG
Species Reactivity: Human, Monkey, Mouse, Rat, Dog, Horse, Sheep and Pig. Others not tested.
Immunogen: A synthetic peptide from human calcitonin
Positive Control: Thyroid or medullary CA
Cellular Localization: Cytoplasmic
Recommended for:
- Immunohistochemistry (Formalin/paraffin)
Note:
USA: RUO; Int'l: RUO
Specifications
Specifications
| Antigen | Calcitonin Ab-2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | Ready-to-use for IHC (UVLP) |
| Host Species | Rabbit |
| Immunogen | A synthetic peptide from human calcitonin |
| Quantity | 1 mL |
| Regulatory Status | RUO |
| Research Discipline | Steroids Hormones and Growth Factors |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.