Learn More
Calcitonin (CALCA) Mouse anti-Human, Clone: OTI2F5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF190005

Description
Peptide sequence is CGNLST CMLGTYTQDF NKFHTFPQTA IGVGAP Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Calcitonin is a 32 amino acid peptide which can be demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm. Amyloid deposits within the tumor may also exhibit varying degrees of calcitonin activity.Specifications
Calcitonin (CALCA) | |
Monoclonal | |
Unconjugated | |
CALCA | |
CALCA | |
Mouse | |
Affinity Chromatography | |
RUO | |
796 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2F5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P06881 | |
CALCA | |
Synthetic peptide sequence | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.