Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calcitonin Monoclonal Antibody (24B2)
GREENER_CHOICE

Catalog No. PIMA120291
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA120291 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA120291 Supplier Invitrogen™ Supplier No. MA120291
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This antibody may be used in a sandwich ELISA when paired with MA1-20285 as the probe antibody.

Calcitonin is a 32 amino acid peptide which can be demonstrated in C cells of the normal and hyperplastic thyroid. Staining for calcitonin may be used for the identification of a spectrum of C cell proliferative abnormalities ranging from C cell hyperplasia to invasive tumors. Staining for calcitonin in medullary carcinoma of the thyroid produces a fine granular pattern in the cytoplasm. Amyloid deposits within the tumor may also exhibit varying degrees of calcitonin activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Calcitonin
Applications ELISA, Western Blot
Classification Monoclonal
Clone 24B2
Concentration 2.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene CALCA
Gene Accession No. P01258, P06881
Gene Alias Alpha CGRP; Alpha type CGRP; Alpha-type CGRP; CA; Cal1; CAL6; CALC; CALC1; Calca; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; calcitonin gene related peptide 1; calcitonin gene related peptide I; calcitonin gene related peptide I precursor; Calcitonin gene-related peptide; calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin precursor; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-like; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRPI; CGRP-I; CT; Ctn; katacalcin; katacalcin (KC); KC; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; procalcitonin; RATCAL6
Gene Symbols CALCA
Host Species Mouse
Immunogen Calcitonin conjugated with carrier protein.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 796
Target Species Human
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.