Learn More
Invitrogen™ Calcitonin Monoclonal Antibody (SP17)

Description
Recommended positive controls: Thyroid or Medullary carcinoma.
Specifications
Specifications
| Antigen | Calcitonin |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Monoclonal |
| Clone | SP17 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Tissue culture supernatant with 0.09% sodium azide |
| Gene | CALCA |
| Gene Accession No. | P01258, P06881 |
| Gene Alias | Alpha CGRP; Alpha type CGRP; Alpha-type CGRP; CA; Cal1; CAL6; CALC; CALC1; Calca; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; calcitonin gene related peptide 1; calcitonin gene related peptide I; calcitonin gene related peptide I precursor; Calcitonin gene-related peptide; calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin precursor; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-like; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRPI; CGRP-I; CT; Ctn; katacalcin; katacalcin (KC); KC; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; procalcitonin; RATCAL6 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.