Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcitonin R Antibody (462802), Janelia Fluor™ 646, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4614J
Description
Calcitonin R Monoclonal antibody specifically detects Calcitonin R in Human samples. It is validated for Flow Cytometry, CyTOF, Gene Knock-OutSpecifications
Calcitonin R | |
Monoclonal | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF, Gene Knock-Out | |
462802 | |
Flow Cytometry, CyTOF-ready, Knockout Validated | |
calcitonin receptor, CRT, CTR, CT-R, CTR1 | |
HEK293 human embryonic kidney cell line transfected with human Calcitonin R, Ala25-Ala474, Accession # NP_001733 | |
0.1 mL | |
Cancer, GPCR, Signal Transduction | |
799 | |
Store at 4°C in the dark. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction