Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcitonin Rabbit anti-Human, Mouse, Porcine, Rat, Polyclonal, FabGennix

Catalog No. 502206579
Encompass
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-220-6579 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-220-6579 Supplier Fabgennix Inc Supplier No. CLTN101AP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Calcitonin Polyclonal antibody specifically detects Calcitonin in Human, Mouse, Porcine, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot

Specifications

Antigen Calcitonin
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.5-1.5 mg/mL
Conjugate Unconjugated
Formulation proprietary buffer with 0.5% BSA, 30% glycerol and 0.02% sodium azide; pH 7.4-7.8
Gene CALCA
Gene Accession No. P01256, P01257, P01258, P01259, P06881, P70160
Gene Alias Alpha CGRP; Alpha-type CGRP; CA; Cal1; CAL6; CALC; CALC1; Calca; Calcitonin; calcitonin 1; Calcitonin carboxyl-terminal peptide; calcitonin gene related peptide I; calcitonin gene related peptide I precursor; Calcitonin gene-related peptide; calcitonin gene-related peptide 1; Calcitonin gene-related peptide I; calcitonin precursor; calcitonin related polypeptide alpha; calcitonin/calcitonin-related polypeptide, alpha; calcitonin; calcitonin gene-related peptide 1; calcitonin-like; calcitonin-related polypeptide alpha; CCALCI; CCP; Cgrp; CGRP1; CGRP-1; CGRPI; CGRP-I; CT; Ctn; katacalcin; KC; PCT; PDN-21; prepro-alpha-calcitonin gene-related peptide; procalcitonin; RATCAL6
Host Species Rabbit
Immunogen Synthetic peptide taken within amino acid region 30-80 on human Calcitonin protein.
Purification Method Affinity chromatography
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100579174, 12310, 24241, 796
Target Species Human, Mouse, Pig, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.