Learn More
Invitrogen™ Calcitonin Receptor Monoclonal Antibody (31/01-1H10-4-1-14)

Description
Mouse anti Human calcitonin recptor antibody, clone 31/01-1H10-4-1-14 recognizes the human calcitonin receptor, a membrane protein with seven transmembrane domains that is coupled to G protein messenger systems.
Specifications
Specifications
| Antigen | Calcitonin Receptor |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 31/01-1H10-4-1-14 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with <0.1% sodium azide |
| Gene | CALCR |
| Gene Accession No. | P30988 |
| Gene Alias | C1A/C1B; calcitonin receptor; CALCR; Clr; COPT1; CRT; CT receptor; CTR; CT-R; CTR1; hCTR1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.