Learn More
Invitrogen™ Calcitonin Receptor Polyclonal Antibody

Description
Calcitonin Receptor Polyclonal Antibody for Western Blot, ICC/IF, Flow
Specifications
Specifications
| Antigen | Calcitonin Receptor |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.09% sodium azide; pH 7.4 |
| Gene | CALCR |
| Gene Accession No. | P30988 |
| Gene Alias | C1A/C1B; calcitonin receptor; CALCR; Clr; COPT1; CRT; CT receptor; CTR; CT-R; CTR1; hCTR1 |
| Gene Symbols | CALCR |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.