Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcitriol 97.0+%, TCI America™
SDP

Catalog No. C30781MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C30781MG 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. C30781MG Supplier TCI America Supplier No. C30781MG

Chemical Identifiers

CAS 32222-06-3
Molecular Formula C27H44O3
Molecular Weight (g/mol) 416.65
MDL Number MFCD00867079,MFCD11656129
InChI Key GMRQFYUYWCNGIN-NKMMMXOESA-N
Synonym calcitriol, rocaltrol, calcijex, topitriol, 1alpha,25-dihydroxyvitamin d3, silkis, soltriol, 1alpha,25-dihydroxycholecalciferol, calcitriolum, dihydroxyvitamin d3
PubChem CID 5280453
ChEBI CHEBI:17823
IUPAC Name (1R,3S,5Z)-5-{2-[(1R,3aS,4E,7aR)-1-[(2R)-6-hydroxy-6-methylheptan-2-yl]-7a-methyl-octahydro-1H-inden-4-ylidene]ethylidene}-4-methylidenecyclohexane-1,3-diol
SMILES C[C@H](CCCC(C)(C)O)[C@H]1CC[C@H]2\C(CCC[C@]12C)=C\C=C1\C[C@@H](O)C[C@H](O)C1=C

Specifications

Melting Point 119°C
Color White
Quantity 1 mg
UN Number 2811
Formula Weight 416.65
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Calcitriol
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.