Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium 3-hydroxy-3-methylbutyrate hydrate, 97+%

Catalog No. AA4272130
Click to view available options
Quantity:
50 g
250 g
1 kg

CAS: 135236-72-5 | C10H18CaO6 | 274.33 g/mol

Calcium 3-hydroxy-3-methylbutyrate hydrate is a nutritional supplement, which represents a calcium salt of beta-hydroxy beta-methylbutyrate (HMB). The latter is a leucine metabolite produced in the body through oxidation of the ketoacid of leucine (ketoisocaproate, KIC). Because leucine plays an important regulatory role in protein metabolism and due to the conversion of leucine to KIC, several researchers have presumed that KIC is the active ingredient responsible for most of the beneficial effects of leucine on protein metabolism.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135236-72-5
Molecular Formula C10H18CaO6
Molecular Weight (g/mol) 274.33
MDL Number MFCD01318562
InChI Key WLJUMPWVUPNXMF-UHFFFAOYSA-L
Synonym calcium 3-hydroxy-3-methyl butyrate
PubChem CID 131847069
SMILES [Ca++].CC(C)(O)CC([O-])=O.CC(C)(O)CC([O-])=O

Specifications

Color White
Quantity 250 g
Solubility Information Soluble in water.
IUPAC Name calcium;3-hydroxy-3-methylbutanoic acid
Formula Weight 274.33 (Anhydrous)
Percent Purity ≥97%
Physical Form Powder
Chemical Name or Material Calcium 3-hydroxy-3-methylbutyrate hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.