Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium, 99%, granular, ACROS Organics™

Click to view available options
Quantity:
5g
100g
500g
Packaging:
Glass bottle
7440-70-2
Ca
40.078
MFCD00085314
OYPRJOBELJOOCE-UHFFFAOYSA-N
unii-sy7q814vup,hsdb 273,sy7q814vup,compounds,elemental,calcium, elemental,kalzium,atom,ingot,chelate
5460341
CHEBI:29320
calcium
[Ca]
7440-70-2
Ca
40.078
MFCD00085314
OYPRJOBELJOOCE-UHFFFAOYSA-N
unii-sy7q814vup,hsdb 273,sy7q814vup,compounds,elemental,calcium, elemental,kalzium,atom,ingot,chelate
5460341
CHEBI:29320
calcium
[Ca]
Quantity 100g
Assay Percent Range 99%
Linear Formula Ca
Fieser 05,89; 09,1634; 14,71
Merck Index 15, 1648
Packaging Glass bottle
Formula Weight 40.07
Physical Form Granules
Percent Purity 99%
Color Gray
Melting Point 845.0°C
Boiling Point 1484.0°C
Chemical Name or Material Calcium, 99%
Show More Show Less

GHS H Statement:
In contact with water releases flammable gases.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Handle under inert gas. Protect from moisture.
Keep away from any possible contact with water, because of violent reaction and possible flash fire.
Brush off loose particles from skin. Immerse in cool water/wrap in wet bandages.
In case of fire: Use CO2, dry chemical, or foam.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 231-179-5

TSCA : TSCA