Calcium, AAS standard solution, Specpure™, Ca 1000µg/mL, Alfa Aesar™

Manufacturer: Alfa Aesar 88057AE

Quantity 100mL

 View more versions of this product

Catalog No. AA88057AE

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7697-37-2,471-34-1,7732-18-5
    Physical Form Liquid
    Molecular Formula CaCO3 in 5% HNO3
    MDL Number MFCD00085314
    Chemical Name or Material Calcium

    Specifications

    Color Clear colorless
    Physical Form Liquid
    Quantity 100mL
    DOT Information DOT Class: 8, Packaging Group: III
    EINECS Number 231-714-2
    Molecular Formula CaCO3 in 5% HNO3
    MDL Number MFCD00085314
    Concentration or Composition (by Analyte or Components) Nitric acid: 5%; Calcium carbonate: 0.25%; Water: 94.75%
    Health Hazard 1 H314: Causes severe skin burns and eye damage.
    CAS 471-34-1,7697-37-2,7732-18-5
    Vapor Pressure 17mm Hg at 20°C
    Chemical Name or Material Calcium

    H314: Causes severe skin burns and eye damage.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit