Alfa Aesar™ Calcium acetate hydrate, Puratronic™, 99.9965% (metals basis)

Manufacturer: Alfa Aesar™ 12974-06

Quantity 5g

 View more versions of this product

Catalog No. AA1297406

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 114460-21-8
    Formula Weight 158.17g/mol (Anhydrous)
    MDL Number MFCD00012448
    Beilstein 3692527
    Merck Index 14,1646
    Chemical Name or Material Calcium acetate hydrate
    Grade Puratronic™

    Specifications

    Density 1.5g/mL
    Sensitivity Hygroscopic
    Melting Point (dec.)
    Quantity 5g
    CAS 114460-21-8
    Physical Form Powder
    Formula Weight 158.17g/mol (Anhydrous)
    MDL Number MFCD00012448
    Beilstein 3692527
    Merck Index 14,1646
    RTECS Number AF7525000
    TSCA TSCA
    Chemical Name or Material Calcium acetate hydrate
    Assay 99.9965%, (metals basis)
    Grade Puratronic™

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit