Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium Activated Nucleotidase 1/CANT1 Antibody (OTI7F8), Alexa Fluor™ 594, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP272378AF594
Description
Calcium Activated Nucleotidase 1/CANT1 Monoclonal specifically detects Calcium Activated Nucleotidase 1/CANT1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Calcium Activated Nucleotidase 1/CANT1 | |
Monoclonal | |
Alexa Fluor 594 | |
50 mM sodium borate with 0.05% sodium azide | |
CANT1 | |
Recombinant protein fragment corresponding to amino acids 63-401 of human Calcium Activated Nucleotidase 1/CANT1 (NP_620148) produced in E.coli. | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI7F8 | |
Western Blot, Immunohistochemistry | |
Apyrase homolog, calcium activated nucleotidase 1, DBQD, EC 3.6.1.6, Putative MAPK-activating protein PM09, Putative NF-kappa-B-activating protein 107, SCAN1, SCAN-1Ca2+-dependent endoplasmic reticulum nucleoside diphosphatase, SHAPYmicromelic dwarfism with vertebral and metaphyseal abnormalities and advancedcarpotarsal ossification, soluble Ca-activated nucleotidase, isozyme 1, soluble calcium-activated nucleotidase 1, soluble calcium-activated nucleotidase SCAN-1 | |
Mouse | |
0.1 mL | |
124583 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction