Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Ascorbate, Dihydrate, USP, 98-101%, Spectrum™ Chemical
SDP

Catalog No. 18601208
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Poly Bottle
Poly Pail

C1764, 5743-28-2, C12H14CaO12·2H2O

Spectrum™ Chemical Calcium Ascorbate, Dihydrate, USP is a form of vitamin C that is used to treat low levels of vitamin C. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5743-28-2
Molecular Formula C12H18CaO14
Molecular Weight (g/mol) 426.34
InChI Key XBTRVIGNXIDJBU-FWCDDDAWSA-N
IUPAC Name calcium bis((5R)-5-[(1S)-1,2-dihydroxyethyl]-3-hydroxy-2,4-dioxooxolan-3-ide) dihydrate
SMILES O.O.[Ca++].OC[C@H](O)[C@H]1OC(=O)[C-](O)C1=O.OC[C@H](O)[C@H]1OC(=O)[C-](O)C1=O

Specifications

pH 6.8-7.4
CAS Max % 100%
Quantity 100 g
Percent Purity 98 to 101%
Grade USP
Loss on Drying 0.1%
Packaging Poly Bottle
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.