Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium benzoate hydrate, 98%, water <8%
Click to view available options
Quantity:
100 g
500 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Calcium benzoate hydrate is used in used in the food industry as a preservative.
Solubility
Soluble in water (2.6 g/100ml at 0°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Calcium benzoate hydrate is used in used in the food industry as a preservative.
Solubility
Soluble in water (2.6 g/100ml at 0°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
Specifications
Specifications
| Molecular Formula | C19H20O4 |
| MDL Number | MFCD00167164 |
| Quantity | 100 g |
| Synonym | bis 4-glycidyloxyphenyl methane, bis 4-glycidyloxy phenyl methane, bis 4-oxiran-2-yl methoxy phenyl methane, para-para-bfdge, p,p-bfdge, diphenylolmethane diglycidyl ether, 2,2'-methylenebis p-phenyleneoxymethylene bisoxirane, bis p-2,3-epoxypropoxy phenyl methane, 4,4'-methylenebisphenol diglycidyl ether, oxirane, 2,2'-methylenebis 4,1-phenyleneoxymethylene bis |
| Solubility Information | Soluble in water (2.6g/100ml at 0°C). |
| InChI Key | XUCHXOAWJMEFLF-UHFFFAOYNA-N |
| SMILES | C1C(O1)COC2=CC=C(C=C2)CC3=CC=C(C=C3)OCC4CO4 |
| IUPAC Name | 2-[[4-[[4-(oxiran-2-ylmethoxy)phenyl]methyl]phenoxy]methyl]oxirane |
| Molecular Weight (g/mol) | 312.37 |
| PubChem CID | 91511 |
| Show More |
Safety and Handling
| EINECSNumber | 218-235-4 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction