Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium binding protein P22 (CHP1) Monoclonal Antibody (OTI3H3), TrueMAB™, OriGene
SDP

Catalog No. 502544033
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4033 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4033 Supplier OriGene Supplier No. CF810154
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Calcium binding protein P22 (CHP1) Monoclonal antibody specifically detects Calcium binding protein P22 (CHP1) in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen Calcium binding protein P22 (CHP1)
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3H3
Concentration See Label
Conjugate Unconjugated
Formulation PBS with 8% trehalose and no preservative; pH 7.3
Gene PODXL2
Gene Accession No. Q9NZ53
Gene Alias 1500003O03Rik; AA960066; AI046351; Cahp; calcineurin B homolog; calcineurin B homologous protein 1; Calcineurin B-like protein; calcineurin homologous protein; calcineurin like EF-hand protein 1; calcineurin-like EF hand protein 1; calcineurin-like EF-hand protein 1; calcium binding protein P22; calcium-binding protein CHP; Calcium-binding protein p22; CHP; chp1; Chromo domain-containing protein 1; chromodomain protein Chp1; EF-hand Ca2+-binding protein p22; EF-hand calcium-binding domain-containing protein p22; p22; p24; si:dkey-41n20.1; sid 470; Sid470; Sid470p; SLC9A1 binding protein; SLC9A1BP; SPAC18G6.02c; Unknown (protein for MGC:127971); Unknown (protein for MGC:63904); vac; Wrch2; wu:fc69b04; zgc:63904
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 366-486 of human PODXL2 produced in E.coli.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 50512
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Lyophilized
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.