Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium bis(trifluoromethylsulfonyl)imide

Catalog No. AA4653303
Click to view available options
Quantity:
250 mg
1 g
5 g

CAS: 165324-09-4 | C4CaF12N2O8S4 | 600.349 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 165324-09-4
Molecular Formula C4CaF12N2O8S4
Molecular Weight (g/mol) 600.349
MDL Number MFCD14156019
InChI Key WFABOCFDABTAPE-UHFFFAOYSA-N
Synonym calcium bis trifluoromethylsulfonyl imide, calcium ii bis trifluoromethanesulfonyl imide, calcium ii triflimide, calcium trifluoromethanesulfonimide, calcium bistrifluoromethanesulfonimide, calcium bis trifluoromethylsulfonyl amide, calcium bis bistriflylimide anion, calcium bis trifluoromethyl sulfonyl amide, bis bis trifluoromethylsulfonyl amino calcium, bis trifluoromethanesulfonyl imide calcium ii salt
PubChem CID 46242197
IUPAC Name calcium;bis(trifluoromethylsulfonyl)azanide
SMILES C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.C(F)(F)(F)S(=O)(=O)[N-]S(=O)(=O)C(F)(F)F.[Ca+2]

Specifications

Melting Point >300°C
Linear Formula [(CF3SO2)2N]2Ca
Quantity 1 g
UN Number UN1759
Sensitivity Moisture sensitive
Solubility Information Insoluble in water.
Formula Weight 600.35
Physical Form Solid
Chemical Name or Material Calcium bis (trifluoromethylsulfonyl)imide
Hazard Category H314-H335
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.