Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium bis(trifluoromethylsulfonyl)imide
Click to view available options
Quantity:
250 mg
1 g
5 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | >300°C |
Linear Formula | [(CF3SO2)2N]2Ca |
Quantity | 5 g |
UN Number | UN1759 |
Sensitivity | Moisture sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 600.35 |
Physical Form | Solid |
Chemical Name or Material | Calcium bis (trifluoromethylsulfonyl)imide |
Safety and Handling
missing translation for 'healthHazard1' | H314-H335 |
missing translation for 'healthHazard2' | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
missing translation for 'healthHazard3' | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S. |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Argon |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction