Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium bromide hydrate, 95%, Alfa Aesar™

Manufacturer:  Alfa Aesar 1882036

Quantity 500g

 View more versions of this product

Catalog No. AA1882036


  Check Availability
Add to cart

Specifications Description & Specifications

Chemical Identifiers

71626-99-8
95.0%
199.9
Calcium bromide hydrate
Powder
CaBr2·xH2O
MFCD00149608

Specifications

71626-99-8
95.0%
White powder
MFCD00149608
White
Calcium bromide hydrate
Powder
CaBr2·xH2O
199.9
Odorless
500g

Safety and Handling

SDS & Safety Documents

H315-H319-H335

EINECSNumber : 232-164-6

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit