Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium carbide

Catalog No. p-4407312
Click to view available options
Quantity:
1 kg
5 x 1 kg

CAS: 75-20-7 | C2Ca

Calcium carbide is used as a desulfurizer, dehydrant of steel, fuel in steel making, powerful deoxidizer and as a source of acetylene gas. It is used as a starting material for the preparation of calcium cyanamide, ethylene, chloroprene rubber, acetic acid, dicyandiamide and cyanide acetate. It is used in carbide lamps, toy cannons such as the big-bang cannon and bamboo cannon. It is associated with calcium phosphide and used in floating, self-igniting naval signal flares. Further, it is involved in the reduction of copper sulfide to metallic copper.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-20-7
Molecular Formula C2Ca
MDL Number MFCD00010905
Synonym calcium carbide, 3-phenoxypropylene di acetate, glycerol phenyl ether diacetate, 3-phenoxy-1,2-propanediol diacetate, phenylglyceryl ether diacetate, 1-phenyl ether-2,3-diacetate glycerol, 1,2-propanediol, 3-phenoxy-, diacetate, 1-acetyloxy-3-phenoxypropan-2-yl acetate, quartz crystal, 3-phenoxypropane-1,2-diyl diacetate

Specifications

Melting Point 2300°C
Color Gray to Black
Physical Form Lump
Quantity 1 kg
Beilstein 3909011
Formula Weight 64.09
Sensitivity Moisture sensitive
Appearance Grayish black lump
Density 2.22 g/cm3
Chemical Name or Material Calcium carbide
Hazard Category H260-H315-H318-H335
Precautionary Statement P223-P231+P232-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P310-P312-P332+P313-P335+P334-P362-P370-P378aa-P378ac-P501c
DOTInformation DOT Class: 4.3; Packaging Group: I
EINECSNumber 200-848-3
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.