Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium carbonate, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E170, precipitated, 98.5-100.5% (based on anhydrous substance), Solstice

Catalog No. 6001915 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
2.5 kg
6 x 1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-019-15 2.5 kg
60-019-14 1 kg
60-044-710 6 x 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-019-15 Supplier Solstice Supplier No. 120102.5KG
Add to Cart
Add to Cart

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E170, precipitated, 98.5-100.5% (based on anhydrous substance)

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Quantity 2.5 kg
Linear Formula CaCO3
UN Number NONH for all modes of transport
Beilstein 8008338
IUPAC Name calcium;carbonate
Formula Weight 100.09g/mol
Percent Purity 98.5 to 100.5% (based on anhydrous substance)
Grade puriss.
Density 2.93g/mL at 25°C
Chemical Name or Material Calcium Carbonate(Caco3), Meets analytical specification of Ph. Eur.BP, USP, FCC
EINECSNumber 207-439-9
RTECSNumber FF9335000
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.