Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Carbonate, Heavy Powder, FCC, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18601214 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-601-214 2.5 kg Poly Pail
18-601-213 500 g Poly Bottle
18-601-215 12 kg Poly Pail
18-601-216 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-601-214 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. C10772.5KG
Add to Cart
Add to Cart

C1077, 471-34-1, CaCO3

Spectrum™ Chemical Calcium Carbonate, Heavy Powder, FCC is used as an acidity regulator, anti-caking agent, preservative and stabilizer in some applications like flour and enriched rice. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food Grade (FCC) chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

CAS Max % 100%
Quantity 2.5 kg
Percent Purity 98 to 100.5%
Grade FCC
Loss on Drying 2%
Packaging Poly Pail
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.