Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Carbonate, Light Powder, USP, 98-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18601230
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
25 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail

CA130, 471-34-1, CaCO3

Spectrum™ Chemical Calcium Carbonate, Light Powder, USP is used as a calcium supplement and as an antacid. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
IUPAC Name calcium carbonate
SMILES [Ca++].[O-]C([O-])=O

Specifications

CAS Max % 100%
Quantity 45 kg
Percent Purity 98 to 100.5%
Grade USP
Loss on Drying 2%
Packaging Fiber Drum
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.