Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium carbonate, Puratronic™, 99.997% (metals basis)

Catalog No. AA4307306
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA4307306 5 g
AA4307314 25 g
AA4307322 100 g
AA4307336 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA4307306 Supplier Thermo Scientific Chemicals Supplier No. 04307306
Add to Cart

CAS: 471-34-1 | CCaO3 | 100.09 g/mol

Calcium carbonate is used in medicine and agriculture. It is also used in flue gas desulfurization in fossil fuel power stations, production of toothpaste, ceramics/glazing applications, purification of iron from iron ore, and to clean tarnish on silver. It is used in the refining of sugar, as filler in thermosetting resins, as an abrasive, as an extender in paints, and as a formation-bridging filtercake-sealing agent in oil industry. It is used in swimming pools for controlling the pH, to improve the rigidity of polypropylene compounds.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 471-34-1
Molecular Formula CCaO3
Molecular Weight (g/mol) 100.09
MDL Number MFCD00010906
InChI Key VTYYLEPIZMXCLO-UHFFFAOYSA-L
Synonym calcium carbonate, limestone, chalk, calcite, carbonic acid calcium salt 1:1, marble, calofort u, aragonite, aeromatt, akadama
PubChem CID 10112
ChEBI CHEBI:3311
SMILES [Ca++].[O-]C([O-])=O

Specifications

Melting Point 800°C (decomposition)
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.997% (Metal basis)
Solubility Information Soluble in acids and insoluble in water.
IUPAC Name calcium carbonate
Formula Weight 100.09
Refractive Index 1.6583
Appearance White powder
Density 2.93
Chemical Name or Material Calcium carbonate
Show More Show Less
EINECSNumber 207-439-9
RTECSNumber FF9335000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.