Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ Calcium Channel beta-4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA597203
Description
Immunogen sequence: ATHTTSSTPM TPLLGRNLGS TALSPYPTAI SGLQSQRMRH SNHSTENSPI ERRSLMTSDE NYHNERARKS RNRLSSSSQH SRDHYPLVEE DYPDSYQDTY KPHRNRGSPG GYSHDSRHRL; Positive Samples: Mouse brain, Rat brain.
This gene encodes a member of the beta subunit family of voltage-dependent calcium channel complex proteins. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. The protein encoded by this locus plays an important role in calcium channel function by modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Certain mutations in this gene have been associated with idiopathic generalized epilepsy and juvenile myoclonic epilepsy. Multiple transcript variants encoding different isoforms have been found for this gene.
Specifications
Calcium Channel beta-4 | |
Polyclonal | |
Unconjugated | |
Cacnb4 | |
3110038O15Rik; CAB4; Cacnb4; CACNLB4; calcium channel beta 4 subunit; calcium channel voltage-dependent subunit beta 4; calcium channel, voltage-dependent, beta 4 subunit; calcium voltage-gated channel auxiliary subunit beta 4; Cchb4; dihydropyridine-sensitive L-type, calcium channel beta-4 subunit; EA5; EIG9; EJM; EJM4; EJM6; lh; OTTHUMP00000207247; OTTHUMP00000207249; Voltage-dependent L-type calcium channel subunit beta-4 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
12298, 58942, 785 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
5.42 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
D4A055, O00305, Q8R0S4 | |
Cacnb4 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 401-520 of human CACNB4 (NP_0007172). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction