Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Calcium Channel beta-4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5110802

Catalog No. PIPA5110802


Only null left
Add to Cart

Description

Description

Immunogen sequence: EHLGEYLEAY WRATHTTSST PMTPLLGRNL GSTALSPYPT AISGLQSQRM RHSNHSTENS PIERRSLMTS DENYHNERAR KSRNRLSSSS QHSRDHYPLV EEDYPDS.

This gene encodes a member of the beta subunit family of voltage-dependent calcium channel complex proteins. Calcium channels mediate the influx of calcium ions into the cell upon membrane polarization and consist of a complex of alpha-1, alpha-2/delta, beta, and gamma subunits in a 1:1:1:1 ratio. Various versions of each of these subunits exist, either expressed from similar genes or the result of alternative splicing. The protein encoded by this locus plays an important role in calcium channel function by modulating G protein inhibition, increasing peak calcium current, controlling the alpha-1 subunit membrane targeting and shifting the voltage dependence of activation and inactivation. Certain mutations in this gene have been associated with idiopathic generalized epilepsy and juvenile myoclonic epilepsy. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Calcium Channel beta-4
Polyclonal
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
O00305
Cacnb4
Recombinant Protein corresonding to Human Calcium Channel beta-4. Recombinant protein control fragment (Product #RP-90685).
100 μL
Primary
Human
Antibody
IgG
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Cacnb4
3110038O15Rik; CAB4; Cacnb4; CACNLB4; calcium channel beta 4 subunit; calcium channel voltage-dependent subunit beta 4; calcium channel, voltage-dependent, beta 4 subunit; calcium voltage-gated channel auxiliary subunit beta 4; Cchb4; dihydropyridine-sensitive L-type, calcium channel beta-4 subunit; EA5; EIG9; EJM; EJM4; EJM6; lh; OTTHUMP00000207247; OTTHUMP00000207249; Voltage-dependent L-type calcium channel subunit beta-4
Rabbit
Antigen Affinity Chromatography
RUO
785
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.