Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium Chloride, 2.75% (w/v), Ricca Chemical
Shop All Ricca Chemical Company Products

Click to view available options
Quantity:
500 mL
1 L
4 L
Packaging:
Natural Poly Bottle
Specifications
Specifications
Color | Colorless |
CAS Min % | 95.0, 3.45 |
CAS Max % | 98.0, 3.59 |
Percent Purity | 2.75% |
Grade | Laboratory |
Packaging | Natural Poly Bottle |
Solvent or Matrix | Aqueous Solution |
pH | 8 to 9 |
Quantity | 4 L |
Physical Form | Liquid |
Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction