Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fisher Science Education™ Calcium Chloride, Anhydrous
SCI_ED

Catalog No. p-4416741 Shop All Fisherbrand Products
Click to view available options
Quantity:
500 g
2 kg
Packaging:
Solid

Catalog No. S25223

Only null left
Add to Cart
Add to Cart

This product is for educational use - no C of A is available.

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
Synonym Dowflake, Caltac
PubChem CID 5284359
ChEBI CHEBI:3312
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

Color Beige
Density Approx 2.15
Material Calcium Chloride
Quantity 500 g
Assay Percent Range >95% w/v
Linear Formula CaCl2
Packaging Solid
Formula Weight 110.99
Chemical Name or Material Calcium Chloride, Anhydrous
ShelfLife 1 year
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.