Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Calcium Chloride, Anhydrous 4 to 8 Mesh, Purified, Reagents
Click to view available options
Quantity:
500 g
3 kg
10 kg
Specifications
Specifications
Boiling Point | 1670°C |
Melting Point | 772°C |
Color | Colorless/White |
pH | 8 to 10 |
Physical Form | Solid |
Quantity | 10 kg |
Grade | Pure |
Packaging | Poly Pail |
Specific Gravity | 2.15 |
Chemical Name or Material | Calcium Chloride, Anhydrous 4 – 8 Mesh, Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction