Alfa Aesar™ Calcium chloride, anhydrous, ACS, 96.0% min

Typical ionic halide, often used as a dessicant

Manufacturer: Alfa Aesar™ 8986618

Quantity 50g

 View more versions of this product

Catalog No. AA8986618

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 10043-52-4
    Molecular Formula CaCl2
    Formula Weight 110.99
    MDL Number MFCD00010903
    Merck Index 14,1659
    Chemical Name or Material Calcium chloride, Anhydrous
    Grade ACS Grade

    Specifications

    Formula Weight 110.99
    Assay Percent Range 96.0% min.
    Molecular Formula CaCl2
    Health Hazard 3 GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.
    MDL Number MFCD00010903
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H319
    Causes serious eye irritation.
    Storage Note 1
    • Packaged under Argon
    Physical Form Powder
    Sensitivity Hygroscopic
    CAS 10043-52-4
    Boiling Point >1600°C
    Melting Point 782°C
    TSCA TSCA
    Merck Index 14,1659
    Quantity 50g
    RTECS Number EV9800000
    Chemical Name or Material Calcium chloride, Anhydrous
    Assay ≥96.0%
    Grade ACS Grade

    GHS H Statement
    H319
    Causes serious eye irritation.

    GHS P Statement
    P280-P264-P305+P351+P338-P337+P313
    Wear protective gloves/protective clothing/eye protection/face protection.
    Wash thoroughly after handling.
    IF IN EYES: Rinse cautiously with water for several minutes.
    Remove contact lenses, if present and easy to do.
    Continue rinsing.
    If eye irritation persists: Get medical advice/attention.

    • Packaged under Argon

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit