Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Anhydrous, ACS Grade, AquaPhoenix Scientific™
SCI_ED

Catalog No. S88518 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S88518 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S88518 Supplier Aqua Phoenix Scientific Supplier No. CC1030J
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 10035-04-8
Molecular Formula CaCl2H4O2
Molecular Weight (g/mol) 147.01
InChI Key LLSDKQJKOVVTOJ-UHFFFAOYSA-L
IUPAC Name calcium dihydrate dichloride
SMILES O.O.[Cl-].[Cl-].[Ca++]

Specifications

CAS Max % 1
Quantity 100 g
Grade ACS
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.