Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Anhydrous, Desiccant, 20 Mesh, Approx. 95%, Spectrum™ Chemical
SDP

Catalog No. 18601233 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
2.5 kg
6 x 500 g
12 kg
Packaging:
Amber Glass Bottle
Poly Pail

Catalog No. 18-601-233

Supplier: Spectrum Chemical Manufacturing Corporation C1090500GMCS6

C1090, 10043-52-4, CaCl2

Spectrum™ Chemical Calcium Chloride, Anhydrous, Desiccant, 20 Mesh is usually a solid white powder with an extremely salty taste. It is highly hygroscopic and must be kept in tightly sealed, air tight containers. Calcium chloride has many uses but its main use is as a desiccant. Ungraded products supplied by Spectrum are indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use. These materials may or may not have a Certificate of Analysis available.

Chemical Identifiers

CAS 10043-52-4
Molecular Formula CaCl2
Molecular Weight (g/mol) 110.98
InChI Key UXVMQQNJUSDDNG-UHFFFAOYSA-L
IUPAC Name calcium dichloride
SMILES [Cl-].[Cl-].[Ca++]

Specifications

CAS Max % 100%
Quantity 6 x 500 g
Packaging Amber Glass Bottle
Percent Purity ∼95%
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.