Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Calcium Chloride, Anhydrous, Flakes / Granules, Technical, Spectrum™

Click to view available options
Quantity:
500 g
2.5 kg
12 kg
100 lb.
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
10043-52-4
10043-52-4
Grade Technical
CAS Max % 1
Packaging Poly Pail
Quantity 12 kg

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.